missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethyl)naphthalene, 97%

Product Code. 11325827
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325827 1 g 1g
11335827 5 g 5g
11315827 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11325827 Supplier Thermo Scientific Alfa Aesar Supplier No. 043491.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13686-49-2 | C12H11Br | 235.124 g/mol

1-(2-Bromoethyl)naphthalene has applications in research field such as Western blotting, Immunoprecipitation, immunogen, and in purification, process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13686-49-2
Molecular Formula C12H11Br
Molecular Weight (g/mol) 235.124
MDL Number MFCD00037737
InChI Key GPHCPUFIWQJZOI-UHFFFAOYSA-N
PubChem CID 139541
IUPAC Name 1-(2-bromoethyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2CCBr

Specifications

Boiling Point 237°C to 238°C (1.65 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Synonym 1-2-bromoethyl naphthalene, 2-1-naphthyl ethyl bromide, naphthalene, 1-2-bromoethyl, 2-bromoethylnaphthalene, 1-2-bromoethyl naphtalene, 1-2-bromo-ethyl-naphthalene, naphthalene,1-2-bromoethyl, 2-alpha-naphthyl ethyl bromide, naphthylathylbromid, 2-bromoethyl napthalene
Solubility Information It is slightly soluble in water.
Formula Weight 235.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1- (2-Bromoethyl)naphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.