missing translation for 'onlineSavingsMsg'
Learn More

1,2-Butylene Oxide, MP Biomedicals

Product Code. 11486455 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486455 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11486455 Supplier MP Biomedicals Supplier No. 0520181180
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 106-88-7
Molecular Formula C4H8O
Quantity 100 g
Synonym 1,2-epoxybutane, 1,2-butylene oxide, oxirane, ethyl, ethyloxirane, 1,2-butene oxide, ethylethylene oxide, 1-butene oxide, oxirane, 2-ethyl, butane, epoxy, 1-butylene oxide
InChI Key RBACIKXCRWGCBB-UHFFFAOYSA-N
SMILES CCC1CO1
IUPAC Name 2-ethyloxirane
Molecular Weight (g/mol) 72.107
PubChem CID 7834
ChEBI CHEBI:82326
Formula Weight 72.11
Chemical Name or Material 1, 2-Butylene Oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.