missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)piperidine hydrochloride, 98%

Product Code. 10676672 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10676672 500 g Plastic bottle 500g
10050860 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10676672 Supplier Thermo Scientific Acros Supplier No. 109215000
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2008-75-5 | C7H14ClN·HCl | 184.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2008-75-5
Molecular Formula C7H14ClN·HCl
Molecular Weight (g/mol) 184.11
MDL Number MFCD00012837
InChI Key VFLQQZCRHPIGJU-UHFFFAOYSA-N
Synonym 1-2-chloroethyl piperidine hydrochloride, 2-piperidinoethylchloride hydrochloride, 2-chloroethylpiperidine hydrochloride, n-chloroethyl piperidine hydrochloride, piperidine, 1-2-chloroethyl-, hydrochloride, 2-piperidinoethyl chloride hydrochloride, piperidinoethyl chloride, hydrochloride, beta-chloroethylpiperidine hydrochloride, 1-2-chloroethyl piperidinium chloride, n-2-chloroethyl piperidine hydrochloride
PubChem CID 74826
IUPAC Name 1-(2-chloroethyl)piperidine;hydrochloride
SMILES C1CCN(CC1)CCCl.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 228°C to 234°C
Color Beige to White
Loss on Drying 1% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 500 g
Beilstein 20, 17
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hot ethanol and hot acetone,fairly soluble in cold ethanol,sparingly soluble in cold acetone,insoluble in benzene and toluene
Formula Weight 184.11
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 1-(2-Chloroethyl)piperidine hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.