missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)piperidine hydrochloride, 98%

Product Code. 11456146
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456146 50 g 50g
11466146 250 g 250g
11404260 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456146 Supplier Thermo Scientific Alfa Aesar Supplier No. B22985.18
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2008-75-5 | C7H15Cl2N | 184.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2008-75-5
Molecular Formula C7H15Cl2N
Molecular Weight (g/mol) 184.104
MDL Number MFCD00012837
InChI Key VFLQQZCRHPIGJU-UHFFFAOYSA-N
Synonym 1-2-chloroethyl piperidine hydrochloride, 2-piperidinoethylchloride hydrochloride, 2-chloroethylpiperidine hydrochloride, n-chloroethyl piperidine hydrochloride, piperidine, 1-2-chloroethyl-, hydrochloride, 2-piperidinoethyl chloride hydrochloride, piperidinoethyl chloride, hydrochloride, beta-chloroethylpiperidine hydrochloride, 1-2-chloroethyl piperidinium chloride, n-2-chloroethyl piperidine hydrochloride
PubChem CID 74826
IUPAC Name 1-(2-chloroethyl)piperidine;hydrochloride
SMILES C1CCN(CC1)CCCl.Cl

Specifications

Melting Point 229°C to 235°C
Quantity 50 g
UN Number UN2928
Beilstein 3910268
Sensitivity Hygroscopic
Formula Weight 184.11
Percent Purity 98%
Chemical Name or Material 1-(2-Chloroethyl)piperidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.