missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chlorophenyl)ethanol, 96%

Product Code. 11426546
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426546 5 g 5g
11436546 25 g 25g
11446546 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426546 Supplier Thermo Scientific Alfa Aesar Supplier No. B24800.06
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 13524-04-4 | C8H9ClO | 156.609 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13524-04-4
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00041037
InChI Key DDUBOVLGCYUYFX-UHFFFAOYSA-N
Synonym 1-2-chlorophenyl ethanol, 1-2-chlorophenyl ethan-1-ol, 2-chloro-alpha-methylbenzyl alcohol, 1-2-chlorophenyl-1-ethanol, benzenemethanol, 2-chloro-.alpha.-methyl, benzenemethanol, 2-chloro-alpha-methyl, benzyl alcohol, o-chloro-.alpha.-methyl, 1-2-chloro-phenyl-ethanol, 1-2-chlorophenyl ethyl alcohol, 1-2'-chlorophenyl-1-hydroxyethane
PubChem CID 26082
IUPAC Name 1-(2-chlorophenyl)ethanol
SMILES CC(C1=CC=CC=C1Cl)O

Specifications

Quantity 5 g
Refractive Index 1.545
Boiling Point 121°C to 123°C (14 mmHg)
Formula Weight 156.61
Percent Purity 96%
Chemical Name or Material 1-(2-Chlorophenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.