missing translation for 'onlineSavingsMsg'
Learn More

1,2-Cyclohexanedicarboxylic anhydride, cis + trans, 97%

Product Code. 11394376
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394376 100 g 100g
11304386 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394376 Supplier Thermo Scientific Alfa Aesar Supplier No. L20001.22
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 85-42-7 | C8H10O3 | 154.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-42-7
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00064863,MFCD00674195
InChI Key MUTGBJKUEZFXGO-UHFFFAOYNA-N
Synonym hexahydrophthalic anhydride, hexahydroisobenzofuran-1,3-dione, hhpa, 1,2-cyclohexanedicarboxylic anhydride, lekutherm hardener h, hexahydrophthalic acid anhydride, 1,3-isobenzofurandione, hexahydro, araldite ht 907, cyclohexane-1,2-dicarboxylic anhydride, 1,2-cyclohexanedicarboxylic acid anhydride
PubChem CID 85689
ChEBI CHEBI:103210
IUPAC Name 3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2CCCCC12

Specifications

Melting Point 32°C to 34°C
Boiling Point 158°C (17 mmHg)
Flash Point 275°C (527°F)
Quantity 100 g
Beilstein 83213
Sensitivity Moisture sensitive
Formula Weight 154.17
Percent Purity 97%
Chemical Name or Material 1,2-Cyclohexanedicarboxylic anhydride, Cis + Trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.