missing translation for 'onlineSavingsMsg'
Learn More

1,2-Cyclohexanediol, cis + trans, 98%

Product Code. 11488497
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488497 10 g 10g
11498497 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488497 Supplier Thermo Scientific Alfa Aesar Supplier No. A18572.09
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 931-17-9 | C6H12O2 | 116.16 g/mol

1,2-Cyclohexanediol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Cyclohexanediol is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931-17-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00003861
InChI Key PFURGBBHAOXLIO-UHFFFAOYSA-N
Synonym 1,2-cyclohexanediol, 2-hydroxycyclohexanol, pyrocatechitol, 1,2-dihydroxycyclohexane, brenzkatechin, 1,2-benzenediol, hexahydro, brenzkatechin german, 1,2-trans-cyclohexanediol, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, mixture of cis and trans
PubChem CID 13601
ChEBI CHEBI:24567
IUPAC Name cyclohexane-1,2-diol
SMILES C1CCC(C(C1)O)O

Specifications

Melting Point 72°C to 76°C
Boiling Point 118°C to 120°C (10 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
Solubility Information Soluble in water.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 1,2-Cyclohexanediol, cis + trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.