missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dehydro Reticuline Iodide, TRC

Product Code. 30366957
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366957 25 mg 25mg
30361605 5 mg 5mg
30352509 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366957 Supplier Toronto Research Chemicals Supplier No. TRCD23006525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21411-21-2
Molecular Formula C19H22INO4
Molecular Weight (g/mol) 455.29
Synonym 3,4-Dihydro-7-hydroxy-1-(3-hydroxy-4-methoxybenzyl)-6-methoxy-2-methylisoquinolinium Iodide, 1,2-Dehydroreticuline Iodide, 3,4-Dihydro-7-hydroxy-1-[(3-hydroxy-4-methoxyphenyl)methyl]-6-methoxy-2-methylisoquinolinium Iodide
SMILES C[N+]1=C(C2=CC(=C(C=C2CC1)OC)O)CC3=CC(=C(C=C3)OC)O.[I-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H21NO4.HI/c1-20-7-6-13-10-19(24-3)17(22)11-14(13)15(20)8-12-4-5-18(23-2)16(21)9-12;/h4-5,9-11H,6-8H2,1-3H3,(H-,21,22);1H
Formula Weight 455.0594
Chemical Name or Material 1,2-Dehydro Reticuline Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.