missing translation for 'onlineSavingsMsg'
Learn More

1,2-Di(2-Aminoethoxy)Ethane-N,N,N,N-Tetraacetic Acid, 98+%, Fisher Chemical

Product Code. 10325990 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10325990 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10325990 Supplier Fisher Chemical Supplier No. D/0755/46
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

25g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

CAS 67-42-5
Melting Point 240°C
Color White
Packaging Plastic powder jar
Molecular Formula C14H24N2O10
MDL Number 4291
Quantity 25 g
Formula Weight 380.34g/mol
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.