missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diaminocyclohexanetetraacetic acid disodium salt solution, Honeywell Fluka™

Product Code. 15618390 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
5 L
6 x 1 L
Unit Size:
1 L
5 L
Pack of 6
Product Code. Quantity unitSize
15618390 1 L 1 L
15624810 5 L 5 L
15638390 6 x 1 L Pack of 6
3 options
This item is not returnable. View return policy

Product Code. 15618390

Brand: Honeywell-Fluka 351031L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric, Reag. Ph. Eur., 0.1 M DCTA-Na4, for complexometry

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5786-78-7
Molecular Formula C14H20N2Na2O8
Molecular Weight (g/mol) 390.3
InChI Key TVTLKUMLAQUFAS-UHFFFAOYSA-L
Synonym sodium 1,2-diaminocyclohexane tetraacetate, 1,2-diaminocyclohexenetetra-acetate of sodium, acetic acid, 1,2-cyclohexylenedinitrilo tetra-, sodium salt, glycine, n,n'-1,2-cyclohexanediylbis n-carboxymethyl-, disodium salt, sodium n'-1,2-cyclohexanediylbis n-carboxymethyl glycinate, glycine, n,n'-1,2-cyclohexanediylbis n-carboxymethyl-, sodium salt 1:2, cdta disodium salt, disodium 2-carboxymethyl-2-carboxymethyl-2-oxido-2-oxoethyl amino cyclohexyl amino acetate
PubChem CID 24230
IUPAC Name disodium;2-[[2-[carboxylatomethyl(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetate
SMILES C1CCC(C(C1)N(CC(=O)O)CC(=O)[O-])N(CC(=O)O)CC(=O)[O-].[Na+].[Na+]

Specifications

Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 6884761
Formula Weight 390.3g/mol
Chemical Name or Material 1,2-Diaminocyclohexanetetraacetic acid disodium salt solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.