missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-1,2-diphenylethane, 96%, Thermo Scientific Chemicals

Product Code. 10245050 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10245050 5 g Glass bottle 5g
10055733 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10245050 Supplier Thermo Scientific Acros Supplier No. 147950050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5789-30-0
Molecular Formula C14H12Br2
Molecular Weight (g/mol) 340.05
MDL Number MFCD00000137
InChI Key GKESIQQTGWVOLH-UHFFFAOYSA-N
Synonym 1,2-dibromo-1,2-diphenylethane, stilbene dibromide, 1,2-dibromo-2-phenylethyl benzene, benzene, 1,1'-1,2-dibromo-1,2-ethanediyl bis, meso-dibromostilbene, benzene, 1,1'-1,2-dibromo-1,2-ethanediyl bis-, r*,s*, alpha,alpha-dibromobibenzyl, bibenzyl,, a,, a'-dibromo, bibenzyl, alpha,alpha'-dibromo, acmc-1ao3z
PubChem CID 93010
IUPAC Name (1,2-dibromo-2-phenylethyl)benzene
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)Br)Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 235°C to 241°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH(Br)CH(Br)C6H5
Quantity 5 g
Beilstein 05, 602
Formula Weight 340.05
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1, 2-Dibromo-1, 2-diphenylethane
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Dibromo-1,2-diphenylethane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.