missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-4-(1,2-dibromoethyl)cyclohexane, TRC

Product Code. 30137905
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137905 10 g 10g
30137393 1 g 1g
30004601 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137905 Supplier Toronto Research Chemicals Supplier No. TRCD42533010G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3322-93-8
Molecular Formula C8 H12 Br4
Molecular Weight (g/mol) 427.8
Synonym 1,2-Dibromo-4-(1,2-dibromoethyl)cyclohexane, 1-(1,2-Dibromoethyl)-3,4-dibromocyclohexane, 4-(1,2-Dibromoethyl)-1,2-dibromocyclohexane, Citex BCL 462, Saytex BCL 462
IUPAC Name 1,2-dibromo-4-(1,2-dibromoethyl)cyclohexane
SMILES BrCC(Br)C1CCC(Br)C(Br)C1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H12Br4/c9-4-8(12)5-1-2-6(10)7(11)3-5/h5-8H,1-4H2
Formula Weight 423.7673
Chemical Name or Material 1,2-Dibromo-4-(1,2-dibromoethyl)cyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.