missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromobutane, 98%

Product Code. 11494247
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494247 50 g 50g
11404257 250 g 250g
11484247 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494247 Supplier Thermo Scientific Alfa Aesar Supplier No. B20356.18
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 533-98-2 | C4H8Br2 | 215.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-98-2
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.92
MDL Number MFCD00000157
InChI Key CZWSZZHGSNZRMW-UHFFFAOYNA-N
Synonym butane, 1,2-dibromo, .alpha.-butylene dibromide, alpha-butylene dibromide, 1,2-dibomobutane, 1,2 dibromobutane, butane,2-dibromo, 1,2-dibromo-butane, alpha-butylene bromide, 1,2-butylene bromide, .alpha.-butylene bromide
PubChem CID 10792
IUPAC Name 1,2-dibromobutane
SMILES CCC(Br)CBr

Specifications

Melting Point -65°C
Density 1.789
Boiling Point 166°C to 167°C
Flash Point >110°C (230°F)
Refractive Index 1.5141
Quantity 50 g
Beilstein 1731380
Merck Index 14,1566
Formula Weight 215.93
Percent Purity 98%
Chemical Name or Material 1,2-Dibromobutane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.