missing translation for 'onlineSavingsMsg'
Learn More

(1,2-Dibromoethyl)benzene, 97%

Product Code. 10267220 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10267220 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10267220 Supplier Thermo Scientific Acros Supplier No. 299305000
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 93-52-7 | C8H8Br2 | 263.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 93-52-7
CAS Min % 96.0
CAS Max % 100.0
Melting Point 68.0°C to 73.0°C
Color Brown to Yellow
Density 1.74g/mL
Boiling Point 139.0°C to 141.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Molecular Formula C8H8Br2
MDL Number MFCD00000138
Quantity 500 g
Beilstein 05, 356
Synonym 1,2-dibromoethyl benzene, dowspray 9, 1,2-dibromo-1-phenylethane, 1,2-dibromo phenyl ethane, benzene, 1,2-dibromoethyl, 1,2-dibromo-2-phenylethane, vic-styrene dibromide, alpha,beta-dibromoethylbenzene, ccris 2090, .alpha.,.beta.-dibromoethylbenzene
InChI Key SHKKTLSDGJRCTR-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(CBr)Br
IUPAC Name 1,2-dibromoethylbenzene
Molecular Weight (g/mol) 263.95
PubChem CID 7145
Formula Weight 263.95
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (1, 2-Dibromoethyl)benzene, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.