missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromopropane, 98%, Thermo Scientific Chemicals

Product Code. 11406166
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406166 500 g 500g
11501346 - -
11496156 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11406166 Supplier Thermo Scientific Alfa Aesar Supplier No. B22994.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1,2-Dibromopropane was used to study the effect of reductant concentration, reductant contact time and suspension pH on reductive dechlorination of carbon tetrachloride by soil manipulated with Fe(II) and HS-. It was used as internal standard during the determination of nitrogenous disinfection byproduct trichloronitromethane by gas chromatography mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dibromopropane was used to study the effect of reductant concentration, reductant contact time and suspension pH on reductive dechlorination of carbon tetrachloride by soil manipulated with Fe(II) and HS-. It was used as internal standard during the determination of nitrogenous disinfection byproduct trichloronitromethane by gas chromatography mass spectrometry.

Solubility
Soluble in water (slightly), ether, and alcohol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-75-1
Molecular Formula C3H6Br2
Molecular Weight (g/mol) 201.889
MDL Number MFCD00000148
InChI Key XFNJYAKDBJUJAJ-UHFFFAOYSA-N
Synonym propylene dibromide, propane, 1,2-dibromo, propylene bromide, 2,3-dibromopropane, ccris 6710, ch2brchbrch3, propane, 1,2-dibromo-, r, propane, 1,2-dibromo-, s, 1,2 dibromopropane, 1,2-dibromo-propane
PubChem CID 6553
IUPAC Name 1,2-dibromopropane
SMILES CC(CBr)Br

Specifications

Melting Point -55°C
Boiling Point 140°C to 142°C
Flash Point 35°C (95°F)
Quantity 500 g
UN Number UN1993
Beilstein 1718884
Merck Index 14,7853
Formula Weight 201.9
Percent Purity 98%
Chemical Name or Material 1,2-Dibromopropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.