missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromotetrachloroethane, TRC

Product Code. 30385025
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30385025 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30385025

Brand: Toronto Research Chemicals TRCD42531525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dibromotetrachloroethane
CAS 630-25-1
Molecular Formula C2Br2Cl4
Quantity 25 g
Synonym 1,2-Dibromo-1,1,2,2-tetrachloroethane, DBTCE, sym-Dibromotetrachloroethane
InChI Formula InChI=1S/C2Br2Cl4/c3-1(5,6)2(4,7)8
SMILES ClC(Cl)(Br)C(Cl)(Cl)Br
IUPAC Name 1,2-dibromo-1,1,2,2-tetrachloroethane
Molecular Weight (g/mol) 325.64
Formula Weight 321.7121
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.