missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-4-nitrobenzene, 99%, Thermo Scientific™

Product Code. 11443324
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443324 250 g 250g
11433324 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443324 Supplier Thermo Scientific Alfa Aesar Supplier No. A10620.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-54-7
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00007207
InChI Key NTBYINQTYWZXLH-UHFFFAOYSA-N
Synonym 3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, dcnb, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, unii-ijt74ji7fs, ccris 3097, 1,2-dichloro-4-nitro-benzene, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech
PubChem CID 7443
IUPAC Name 1,2-dichloro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 39°C to 43°C
Boiling Point 255°C to 256°C
Flash Point 123°C (253°F)
Quantity 250 g
UN Number UN3077
Beilstein 1818163
Formula Weight 192
Percent Purity 99%
Chemical Name or Material 1,2-Dichloro-4-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.