missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, 99%, Reagent Grade, Honeywell™

Product Code. 15617820 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
Unit Size:
1 L
100mL
2.5 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15617820 100 mL Glass bottle 100mL
15627820 1 L Glass bottle 1 L
15637820 2.5 L Glass bottle 2.5 L
15647820 20 L Stainless steel can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15617820 Supplier Honeywell Research Chemicals Supplier No. D56802100ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-50-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000535
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
PubChem CID 7239
ChEBI CHEBI:35290
IUPAC Name 1,2-dichlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Cl

Specifications

Melting Point -17°C
Color Colorless to Yellow
Density 1.306g/mL at 25°C
Boiling Point 180°C (1013 hPa)
Explosion Limit 0.092
Flash Point 66°C (150.8°F)
Ignition Point 1198°F
Odor Aromatic
Packaging Glass bottle
Quantity 100 mL
UN Number UN1591
Beilstein 606078
Vapor Density 5.1 (vs air)
Formula Weight 147g/mol
Vapor Pressure 1.6mmHg (35°C), 1.2mmHg (20°C)
Percent Purity 0.99
Chemical Name or Material 1,2-Dichlorobenzene
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.