missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, 99+%, for spectroscopy

Product Code. 10657893 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10657893 2.5 L Glass bottle 2.5 L
10551471 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10657893 Supplier Thermo Scientific Acros Supplier No. 222050025
  • ONLINE EXCLUSIVE PRICE
    274.00 €
    266.00 € / 2.5 L
    Save 8.00 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 95-50-1 | C6H4Cl2 | 147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-50-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 147
MDL Number MFCD00000535
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
PubChem CID 7239
ChEBI CHEBI:35290
IUPAC Name 1,2-dichlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Cl

Specifications

Alkalinity 0.0005 meq/g max.
Acidity 0.0003 meq/g max.
Absorbance (1 cm cell vs water), 0.01 at 380nm, 0.05 at 336nm, 0.3 at 300nm, 1 at 296nm
CAS Min % 99.0
CAS Max % 100.0
Melting Point -15.0°C
Density 1.3000g/mL
Boiling Point 179.0°C to 180.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H4Cl2
Refractive Index 1.5500 to 1.5520
Quantity 2.5 L
Beilstein 05, 201
Residue after Evaporation 0.0005% max.
Merck Index 15, 3065
Specific Gravity 1.3
Solubility Information Solubility in water: 0.13g/L(20°C). Other solubilities: miscible with most common organic solvents,miscible with alcohol,ether and benzene
Formula Weight 147
Percent Purity 99+%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material 1, 2-Dichlorobenzene, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.