missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, CHROMASOLV™, 99%, Honeywell Riedel-de Haen

Product Code. 15622860
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
Packaging:
Glass Bottle
missing translation for 'unitSize'
1 L
This item is not returnable. View return policy

Product Code. 15622860

missing translation for 'mfr': Honeywell-Chromasolv 2705981L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CHROMASOLV™, 99%

  • High purity chromatography solvents
  • Suitability tested for specified use from HPLC up to UHPLC-MS applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 95-50-1
Absorbance max. 1.0 at 296 nm, max. 0.01 at 400 nm, max. 0.01 at 375 nm, max. 0.05 at 335 nm, max. 0.20 at 305 nm, max. 0.30 at 300 nm
CAS Max % 100.0000%
Melting Point -18°C to -17°C (literature)
Density 1.306 g/mL at 25 °C(lit.)
Boiling Point 180°C
Flash Point 66°C
Packaging Glass Bottle
Molecular Formula C6H4Cl2
Linear Formula C6H4Cl2
MDL Number MFCD00000535
Quantity 1 L
UN Number UN1591
Beilstein 606078
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)Cl)Cl
IUPAC Name 1,2-dichlorobenzene
Molecular Weight (g/mol) 146.998
PubChem CID 7239
ChEBI CHEBI:35290
Formula Weight 147
Percent Purity >98.5%
Grade High Purity
Chemical Name or Material 1,2-Dichlorobenzene, CHROMASOLV™, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.