missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene-d4, for NMR, 98 atom% D

Product Code. 10388980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10388980 1 g Glass Bottle 1g
10239893 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10388980 Supplier Thermo Scientific Acros Supplier No. 321260010
  • ONLINE EXCLUSIVE PRICE
    93.65 €
    90.90 € / 1g
    Save 2.75 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 2199-69-1 | C6H4Cl2 | 151.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2199-69-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 151.02
MDL Number MFCD00037106
InChI Key RFFLAFLAYFXFSW-RHQRLBAQSA-N
Synonym 1,2-dichlorobenzene-d4, 1,2-dichlorobenzene d4, benzene-1,2,3,4-d4-, 5,6-dichloro, o-dichloro 2h4 benzene, deuterated 1,2-dichlorobenzene, tetradeutero-1,2-dichlorobenzene, 1,2-dichlorobenzene-d4, 98 atom % d, 1,2-dichlorobenzene-d4,98atom%d, benzene-1,2,3,4-d4, 5,6-dichloro, 1,2-dichloro-3,4,5,6-tetradeuterobenzene
PubChem CID 519913
IUPAC Name 1,2-dichloro-3,4,5,6-tetradeuteriobenzene
SMILES [2H]C1=C([2H])C([2H])=C(Cl)C(Cl)=C1[2H]

Specifications

Physical Form Liquid
Color Colorless
Density 1.3400g/mL
Deuterium (D) 97.5% min.
Packaging Glass Bottle
Water 0.2% max. (Coulometric)
Quantity 1 g
Linear Formula C6D4Cl2
Formula Weight 151.04
Grade NMR
Chemical Name or Material 1, 2-Dichlorobenzene-d4, 98 atom% D
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.