missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene-d4, for NMR, 99 atom % D

Product Code. 16328206 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16328206 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16328206 Supplier Thermo Scientific Acros Supplier No. 464060010
  • ONLINE EXCLUSIVE PRICE
    113.00 €
    103.00 € / 1mL
    Save 10.00 €
    8% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2199-69-1 | C6H4Cl2 | 151.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.3400 g/mL
Flash Point 65°C
Refractive Index 1.5509
Specific Gravity 1.34
Quantity 1 mL
CAS 2199-69-1
Molecular Formula C6H4Cl2
Linear Formula C6D4Cl2
MDL Number MFCD00037106
Solubility Information Solubility aq. soln.: 0.13 g/L water (20°C). Other solubilities: miscible with most common organic solvents, miscible with alcohol, ether and benzene
InChI Key RFFLAFLAYFXFSW-RHQRLBAQSA-N
SMILES [2H]C1=C([2H])C([2H])=C(Cl)C(Cl)=C1[2H]
IUPAC Name dichloro(²H₄)benzene
Molecular Weight (g/mol) 151.02
Formula Weight 151.04
Grade NMR
Chemical Name or Material 1,2-Dichlorobenzene-d4
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.