missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haen

Catalog Number 15624650
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
107-06-2
C2H4Cl2
98.95
MFCD00000963
WSLDOOZREJYCGB-UHFFFAOYSA-N
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
11
CHEBI:27789
1,2-dichloroethane
ClCCCl
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
107-06-2
C2H4Cl2
98.95
MFCD00000963
WSLDOOZREJYCGB-UHFFFAOYSA-N
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
11
CHEBI:27789
1,2-dichloroethane
ClCCCl

CHROMASOLV™, for HPLC,≥99.8%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY
Melting Point -35°C (literature)
Absorbance max. 0.01 at 270 nm, max. 0.04 at 245 nm, max. 0.60 at 230 nm
CAS Max % ≤100.0000%
Linear Formula ClCH2CH2Cl
UN Number UN1184
Beilstein 605264
Formula Weight 98.96
Percent Purity >99.8%
Grade HPLC
Density 1.256 g/mL at 25 °C(lit.)
Boiling Point 83°C
Quantity 1 L
Flash Point 13°C
Packaging Glass Bottle
Chemical Name or Material 1,2-Dichloroethane, CHROMASOLV™, for HPLC, ≥99.8%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'