missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane-d{4}, 99% (Isotopic)

Product Code. 11386069
Change view
Click to view available options
Quantity:
1 g
5 g
5 x 1 g
Unit Size:
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386069 5 x 1 g 5g
11369967 1 g 1g
11379967 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11386069 Supplier Thermo Scientific Alfa Aesar Supplier No. 042281.46
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 17060-07-0 | C2H4Cl2 | 102.978 g/mol

1,2-Dichloroethane-d4 is widely used as a solvent for NMR analysis, and in the synthesis of poly(ethylene-d4 oxide) as a chain termination reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17060-07-0
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 102.978
MDL Number MFCD00037531
InChI Key WSLDOOZREJYCGB-LNLMKGTHSA-N
Synonym 1,2-dichloroethane-d4, 1,2-dichloroethane d4, ethylene-d4 dichloride, dichloro 2 h? ethane, 1,2-dichloroethane-d4, 99 atom % d, 1,2-dichloroethane-d4 98.0 atom % d, 1,2-dichloroethane-d4, analytical standard, 1,2-dichloroethane d4 2000 ng/microl in methanol
PubChem CID 12197860
IUPAC Name 1,2-dichloro-1,1,2,2-tetradeuterioethane
SMILES C(CCl)Cl

Specifications

Physical Form Liquid
Density 1.307 g/mL
Flash Point 15°C (60°F)
Refractive Index 1.443
Quantity 5 x 1 g
Melting Point -35°C
Boiling Point 83°C
UN Number UN1184
Solubility Information Partially miscible with water.
Formula Weight 102.99
Percent Purity 99%
Chemical Name or Material 1,2-Dichloroethane-d4, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.