missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diethoxyethane, 98%

Product Code. 11329085
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11329085

Brand: Thermo Scientific Alfa Aesar L14283.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 629-14-1 | C6H14O2 | 118.176 g/mol

Ethylene glycol diethyl ether is a important solvent, used for ink, paint and coating industry. Mainly used in polymer, electrochemistry, boracium chemistry, resin, nitro cellulose, In this product , the benzene, xenene, terphenyl, anthracene reacts with sodium and then bring about the complex; it is also used in surface treatment, halogen analysis in gasoline, acetic acid recycle in diluted acetic acid, depaint agent, thinner, flushing agent, stabilizer, anti oxidant , thickener of lube.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene glycol diethyl ether is a important solvent, used for ink, paint and coating industry. Mainly used in polymer, electrochemistry, boracium chemistry, resin, nitro cellulose, In this product , the benzene, xenene, terphenyl, anthracene reacts with sodium and then bring about the complex; it is also used in surface treatment, halogen analysis in gasoline, acetic acid recycle in diluted acetic acid, depaint agent, thinner, flushing agent, stabilizer, anti oxidant , thickener of lube.

Solubility
Soluble in water 34 g/L @ 20˚C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-14-1
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00009253
InChI Key LZDKZFUFMNSQCJ-UHFFFAOYSA-N
Synonym ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu
PubChem CID 12375
IUPAC Name 1,2-diethoxyethane
SMILES CCOCCOCC

Specifications

Melting Point -74°C
Density 0.842
Boiling Point 120°C to 121°C
Flash Point 19°C (66°F)
Linear Formula CH3CH2OCH2CH2OCH2CH3
Refractive Index 1.392
Quantity 500 mL
UN Number UN1153
Beilstein 1732917
Solubility Information Soluble in water 34g/L @ 20?C.
Formula Weight 118.18
Percent Purity 98%
Chemical Name or Material 1,2-Diethoxyethane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.