missing translation for 'onlineSavingsMsg'
Learn More

1,2-Difluoro-4-nitrobenzene, 98+%

Product Code. 11428913
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428913 50 g 50g
11418913 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428913 Supplier Thermo Scientific Alfa Aesar Supplier No. A11060.18
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 369-34-6 | C6H3F2NO2 | 159.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-34-6
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.092
MDL Number MFCD00007198
InChI Key RUBQQRMAWLSCCJ-UHFFFAOYSA-N
Synonym 3,4-difluoronitrobenzene, benzene, 1,2-difluoro-4-nitro, 1-nitro-3,4-difluorobenzene, 1,2-difluoro-4-nitro-benzene, 3,4-difluoro nitrobenzene, pubchem2290, 3,4-difluornitrobenzene, 3.4-difluoronitrobenzene, 3, 4-difluoronitrobenzene, 3,4-difluoro-nitrobenzene
PubChem CID 123053
IUPAC Name 1,2-difluoro-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])F)F

Specifications

Density 1.441
Boiling Point 76°C to 80°C (8 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.508
Quantity 50 g
UN Number UN2810
Beilstein 1944996
Formula Weight 159.09
Percent Purity ≥98%
Chemical Name or Material 1,2-Difluoro-4-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.