missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dihydroxybenzene-3,5-Disulfonic Acid, Disodium Salt, Pure, Metal Indicator, Fisher Chemical

Product Code. 10448210 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10448210 25 g Plastic powder jar 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10448210 Supplier Fisher Chemical Supplier No. D/3000/46
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 300°C
CAS 149-45-1
Color White
Physical Form Powder/Solid
Quantity 25 g
Molecular Formula C6H10Na2O8S2
MDL Number 149531
Synonym tiron, tiferron, sodium catechol sulfate, disodium 4,5-dihydroxybenzene-1,3-disulfonate, 3,5-disulfocatechol disodium salt, sodium pyrocatechol-3,5-disulfonate, disodium pyrocatechol-3,5-disulfonate, sodium 1,2-dihydroxybenzenedisulfonate, 1,2-dihydroxybenzene-3,5-disulfonic acid disodium salt, 1,3-benzenedisulfonic acid, 4,5-dihydroxy-, disodium salt
InChI Key HEOKHLCODUWALT-UHFFFAOYSA-N
SMILES [HH].[HH].C1=C(C=C(C(=C1S(=O)(=O)O)O)O)S(=O)(=O)O.[Na].[Na]
IUPAC Name 4,5-dihydroxybenzene-1,3-disulfonic acid;molecular hydrogen;sodium
Molecular Weight (g/mol) 320.238
PubChem CID 131674010
Formula Weight 332.21g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.