missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethoxyethane, CHROMASOLV™, for HPLC, 99.9%, inhibitor-free, Honeywell Riedel-de Haën™

Product Code. 15653120
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15653120 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15653120 Supplier Honeywell-Chromasolv Supplier No. 3074321L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 110-71-4
CAS Max % ≤100.0000%
Melting Point -58°C (literature)
Density 0.867 g/mL at 25 °C(lit.)
Boiling Point 85°C
Flash Point −2°C
Packaging Glass Bottle
Molecular Formula C4H10O2
Linear Formula CH3OCH2CH2OCH3
MDL Number MFCD00008502
Quantity 1 L
UN Number UN2252
Beilstein 1209237
Synonym monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether
InChI Key XTHFKEDIFFGKHM-UHFFFAOYSA-N
SMILES COCCOC
IUPAC Name 1,2-dimethoxyethane
Molecular Weight (g/mol) 90.122
PubChem CID 8071
ChEBI CHEBI:42263
Formula Weight 90.12
Percent Purity >99.85%
Grade HPLC
Chemical Name or Material 1,2-Dimethoxyethane, CHROMASOLV™, for HPLC, 99.9%, inhibitor-free
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,2-Dimethoxyethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H360FD-May damage fertility. May damage the unborn child.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-Dimethoxyethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.