missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethoxyethane, TRC

Product Code. 30387700
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387700 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387700 Supplier Toronto Research Chemicals Supplier No. TRCD460625100ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dimethoxyethane
CAS 110-71-4
Purity Grade Notes GC
Molecular Formula C4 H10 O2
Quantity 100 mL
Synonym Ethane, 1,2-dimethoxy-, 1,2-Dimethoxyethane, 1,2-Ethanediol, dimethyl ether, 2,5-Dioxahexane, DME, DME (glycol ether), Dimethyl Cellosolve, Ethylene dimethyl ether, Ethylene glycol dimethyl ether, Glycol dimethyl ether, Glyme, Hisolve MMM, Methyl Monoglyme, Monoethylene glycol dimethyl ether, Monoglyme, NSC 60542, α,β-Dimethoxyethane
InChI Formula InChI=1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3
SMILES COCCOC
IUPAC Name 1,2-dimethoxyethane
Molecular Weight (g/mol) 90.12
Formula Weight 90.0681
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.