missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethyl 4-nitrophthalate, TRC

Product Code. 30384370
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384370 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384370 Supplier Toronto Research Chemicals Supplier No. TRCD480408500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 610-22-0
Molecular Formula C10 H9 N O6
Quantity 500 mg
Synonym 4-Nitrophthalic Acid, Bis-methyl Ester, 1,2-Benzenedicarboxylic acid, 4-nitro-, dimethyl ester (9CI), Phthalic acid, 4-nitro-, dimethyl ester (6CI,7CI,8CI), 1,2-Bis(methoxycarbonyl)-4-nitrobenzene, 1,2-Dimethyl 4-nitrobenzene-1,2-dicarboxylate, 4-Nitro-1,2-benzenedicarboxylic acid dimethyl ester, 4-Nitrophthalic acid dimethyl ester, Dimethyl 4-nitrobenzene-1,2-dicarboxylate, Dimethyl 4-nitrophthalate, NSC 9410, 1,2-Benzenedicarboxylic acid, 4-nitro-, 1,2-dimethyl ester
InChI Formula InChI=1S/C10H9NO6/c1-16-9(12)7-4-3-6(11(14)15)5-8(7)10(13)17-2/h3-5H,1-2H3
SMILES COC(=O)c1ccc(cc1C(=O)OC)[N+](=O)[O-]
IUPAC Name dimethyl 4-nitrobenzene-1,2-dicarboxylate
Molecular Weight (g/mol) 239.182
Formula Weight 239.043
Chemical Name or Material 4-Nitrophthalic Acid, Bis-methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.