missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethylnaphthalene, 98%

Product Code. 11312267
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312267 250 mg 250mg
11302267 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312267 Supplier Thermo Scientific Alfa Aesar Supplier No. L09774.MD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 573-98-8 | C12H12 | 156.228 g/mol

1,2-Dimethylnaphthalene is suitable reagent used to investigate the secondary organic aerosol (SOA) production via gas-phase photooxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dimethylnaphthalene is suitable reagent used to investigate the secondary organic aerosol (SOA) production via gas-phase photooxidation.

Solubility
Difficult to mix in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 573-98-8
Molecular Formula C12H12
Molecular Weight (g/mol) 156.228
MDL Number MFCD00004035
InChI Key QNLZIZAQLLYXTC-UHFFFAOYSA-N
Synonym dimethylnaphthalene, naphthalene, dimethyl, naphthalene, 1,2-dimethyl, 1,2-dimethyl naphthalene, unii-23t7o135bd, 1,2-dmn, 1,2-dimethyl-naphthalene, dsstox_cid_5130, dsstox_rid_77675, dsstox_gsid_25130
PubChem CID 11317
ChEBI CHEBI:34052
IUPAC Name 1,2-dimethylnaphthalene
SMILES CC1=C(C2=CC=CC=C2C=C1)C

Specifications

Density 1.013
Boiling Point 266°C to 267°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.616
Quantity 250 mg
Beilstein 2039376
Solubility Information Difficult to mix in water.
Formula Weight 156.23
Percent Purity 98%
Chemical Name or Material 1,2-Dimethylnaphthalene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.