missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Diphenylethane, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 1,2-Diphenylethane |
| CAS | 103-29-7 |
| Molecular Formula | C14 H14 |
| Quantity | 1 g |
| Synonym | Benzene, 1,1'-(1,2-ethanediyl)bis-, Bibenzyl (8CI), 1,1'-(1,2-Ethanediyl)bis[benzene], (2-Phenylethyl)benzene, 1,2-Dihydrostilbene, 1,2-Diphenylethane, Dibenzil, Dibenzyl, Dihydrostilbene, Ethane, 1,2-diphenyl-, Ethylenebis[benzene], NSC 30686, NSC 8789, sym-Diphenylethane, Bibenzyl, Dibenzyl, Dihydrostilbene |
| InChI Formula | InChI=1S/C14H14/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-10H,11-12H2 |
| SMILES | C(Cc1ccccc1)c2ccccc2 |
| IUPAC Name | 2-phenylethylbenzene |
| Molecular Weight (g/mol) | 182.26 |
| Formula Weight | 182.1096 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction