missing translation for 'onlineSavingsMsg'
Learn More

1,2-Distearoyl-sn-glycero-3-phosphoethanolamine, TRC

Product Code. 30387343
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387343 500 mg 500mg
30370346 50 mg -
30371938 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387343 Supplier Toronto Research Chemicals Supplier No. TRCD493610500MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1069-79-0
Molecular Formula C41H82NO8P
Molecular Weight (g/mol) 748.07
Synonym Octadecanoic Acid (1R)-1-[[[(2-Aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl Ester, (R)-Octadecanoic Acid 1-[[[(2-Aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl Ester, L-1,2-Di-stearin 2-Aminoethyl hydrogen phosphate, (R)-Distearoylphosphatidylethanolamine, 1,2-Distearoyl-sn-glycero-3-phosphatidylethanolamine, 1,2-Distearoyl-sn-glycero-3-phosphoethanolamine, DSPE, Distearoyl-L-alpha-phosphatidylethanolamine, L-alpha-Distearoylphosphatidylethanolamine, L-beta,gamma-Distearoyl-alpha-phosphatidylethanolamine
IUPAC Name [(2R)-3-[2-aminoethoxy(hydroxy)phosphoryl]oxy-2-octadecanoyloxypropyl] octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)(O)OCCN)OC(=O)CCCCCCCCCCCCCCCCC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C41H82NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(43)47-37-39(38-49-51(45,46)48-36-35-42)50-41(44)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h39H,3-38,42H2,1-2H3,(H,45,46)/t39-/m1/s1
Formula Weight 747.5778
Chemical Name or Material 1,2-Distearoyl-sn-glycero-3-phosphoethanolamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.