missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxy-3-methylbutane, 98+%

Product Code. 11354176
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354176 1 g 1g
11364176 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354176 Supplier Thermo Scientific Alfa Aesar Supplier No. L19291.03
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 1438-14-8 | C5H10O | 86.134 g/mol

1,2-Epoxy-3-methylbutane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxy-3-methylbutane is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Store under inert gas. It is sensitive to moisture. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1438-14-8
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD02683501
InChI Key REYZXWIIUPKFTI-UHFFFAOYSA-N
Synonym 2-isopropyloxirane, 1,2-epoxy-3-methylbutane, isopropyloxirane, oxirane, 1-methylethyl, butane, 1,2-epoxy-3-methyl, 2-propan-2-yl oxirane, oxirane,2-1-methylethyl, 1-methylethyl-oxirane, isopropylethylene oxide, acmc-1asns
PubChem CID 102618
IUPAC Name 2-propan-2-yloxirane
SMILES CC(C)C1CO1

Specifications

Density 0.815
Boiling Point 72°C to 75°C
Flash Point −12°C (10°F)
Odor Recognizable
Refractive Index 1.3915
Quantity 1 g
UN Number UN1993
Beilstein 102418
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 86.13
Percent Purity ≥98%
Chemical Name or Material 1,2-Epoxy-3-methylbutane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.