missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxydecane, 97%

Product Code. 11317206
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317206 5 g 5g
11327206 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317206 Supplier Thermo Scientific Alfa Aesar Supplier No. L07289.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2404-44-6 | C10H20O | 156.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2404-44-6
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00005158
InChI Key AAMHBRRZYSORSH-UHFFFAOYNA-N
Synonym 1,2-epoxydecane, oxirane, octyl, octyloxirane, oxirane, 2-octyl, 1,2-decylene oxide, ccris 2615, epoxides, c>8-alkyl, decene oxide, epoxy decane, oxirane,octyl
PubChem CID 16993
IUPAC Name 2-octyloxirane
SMILES CCCCCCCCC1CO1

Specifications

Density 0.84
Boiling Point 92°C to 93°C (14 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.429
Quantity 5 g
Beilstein 104132
Formula Weight 156.27
Percent Purity 97%
Chemical Name or Material 1,2-Epoxydecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.