missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxyhexane, 96%

Product Code. 11371977
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371977 25 g 25g
11381977 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371977 Supplier Thermo Scientific Alfa Aesar Supplier No. L07609.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1436-34-6 | C6H12O | 100.161 g/mol

1,2-Epoxyhexane is used as chemical reagent, organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxyhexane is used as chemical reagent, organic intermediates, fine chemicals, pharmaceutical research and development.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1436-34-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00005154
InChI Key WHNBDXQTMPYBAT-UHFFFAOYSA-N
Synonym 1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxide
PubChem CID 15036
IUPAC Name 2-butyloxirane
SMILES CCCCC1CO1

Specifications

Density 0.832
Boiling Point 118°C to 120°C
Flash Point 15°C (59°F)
Refractive Index 1.406
Quantity 25 g
UN Number UN1993
Beilstein 102568
Solubility Information Difficult to mix.
Formula Weight 100.16
Percent Purity 96%
Chemical Name or Material 1,2-Epoxyhexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.