missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxyoctadecane, tech. 85%

Product Code. 11371987
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371987 25 g 25g
11381987 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371987 Supplier Thermo Scientific Alfa Aesar Supplier No. L07622.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7390-81-0 | C18H36O | 268.485 g/mol

1,2-Epoxyoctadecane can be used to produce octadecane-1,2-diol at heating. It is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxyoctadecane can be used to produce octadecane-1,2-diol at heating. It is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Insoluble in water.

Notes
Protect from moisture. Store away from incompatible materials such as oxidizing agents. Moisture-sensitive. Incompatible with strong acids, peroxides, caustics.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7390-81-0
Molecular Formula C18H36O
Molecular Weight (g/mol) 268.485
MDL Number MFCD00005152
InChI Key QBJWYMFTMJFGOL-UHFFFAOYSA-N
Synonym 1,2-epoxyoctadecane, hexadecyloxirane, oxirane, hexadecyl, octadecane, epoxy, 1,2-octadecylene oxide, ccris 2619, oxirane, 2-hexadecyl, #, 1,2-epoxy octadecane, acmc-209osu
PubChem CID 23872
IUPAC Name 2-hexadecyloxirane
SMILES CCCCCCCCCCCCCCCCC1CO1

Specifications

Melting Point 30°C to 35°C
Boiling Point 146°C to 148°C (1 mmHg)
Flash Point 176°C (348°F)
Quantity 25 g
Beilstein 115434
Solubility Information Insoluble in water.
Formula Weight 268.49
Percent Purity 85%
Grade Technical
Chemical Name or Material 1,2-Epoxyoctadecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.