missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxyoctane, 97%

Product Code. 11456984
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456984 25 g 25g
11466984 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456984 Supplier Thermo Scientific Alfa Aesar Supplier No. B21463.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2984-50-1 | C8H16O | 128.22 g/mol

It is employed as an intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate for organic synthesis.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2984-50-1
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00005157
InChI Key NJWSNNWLBMSXQR-UHFFFAOYNA-N
Synonym 1,2-epoxyoctane, hexyloxirane, 1-octene oxide, octylene epoxide, oxirane, hexyl, 1-octene epoxide, octane 1,2-oxide, octene-1,2-oxide, 1,2-epoxy-n-octane, n-octene-1,2-oxide
PubChem CID 18126
IUPAC Name 2-hexyloxirane
SMILES CCCCCCC1CO1

Specifications

Density 0.838
Boiling Point 167°C
Flash Point 37°C (98°F)
Refractive Index 1.42
Quantity 25 g
UN Number UN1993
Beilstein 79912
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material 1,2-Epoxyoctane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.