missing translation for 'onlineSavingsMsg'
Learn More

[1-(2-Furylmethyl)piperid-4-yl]methylamine, 95%, Thermo Scientific™

Product Code. 10515944
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10515944 250 mg Amber Glass Bottle 250mg
10062013 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10515944 Supplier Thermo Scientific Maybridge Supplier No. CC68513CB
 Restricted Item
Estimated Shipment: 12-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 725211-89-2
Molecular Formula C11H18N2O
Molecular Weight (g/mol) 194.28
MDL Number MFCD09837352
InChI Key GKCIOGOJFDDOJI-UHFFFAOYSA-N
Synonym 1-2-furylmethyl piperid-4-yl methylamine, 1-2-furylmethyl piperidin-4-yl methylamine, 1-furan-2-ylmethyl piperidin-4-yl methanamine, 1-1-furan-2-ylmethyl piperidin-4-yl methanamine, 1-2-furylmethyl piperidin-4-yl methyl amine, 1-1-furan-2-yl methyl piperidin-4-yl methanamine
PubChem CID 11816398
SMILES NCC1CCN(CC2=CC=CO2)CC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 1-{1-[(furan-2-yl)methyl]piperidin-4-yl}methanamine
Formula Weight 194.28
Percent Purity 95%
Chemical Name or Material [1-(2-Furylmethyl)piperid-4-yl]methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.