missing translation for 'onlineSavingsMsg'
Learn More

1,2-Hexanediol, 97%

Product Code. 11327026
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327026 250 g 250g
11317026 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327026 Supplier Thermo Scientific Alfa Aesar Supplier No. L06864.30
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6920-22-5 | C6H14O2 | 118.18 g/mol

1,2-Hexanediol is widely used in various cosmetic applications such as hair conditioners, deodorants and antiperspirants. It is involved in the modification of sodium dodecyl sulfate micelles, which is used for amino acid separation by micellar electrokinetic chromatography (MEKC) method. It is a moisturizing agent and plays a vital role in baby care products, bath products, eye makeup, cleansing products and skin care products. It is also used as an additive in liquid cleaning compositions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Hexanediol is widely used in various cosmetic applications such as hair conditioners, deodorants and antiperspirants. It is involved in the modification of sodium dodecyl sulfate micelles, which is used for amino acid separation by micellar electrokinetic chromatography (MEKC) method. It is a moisturizing agent and plays a vital role in baby care products, bath products, eye makeup, cleansing products and skin care products. It is also used as an additive in liquid cleaning compositions.

Solubility
Miscible with water, lower aliphatic hydrocarbons and fatty acids.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6920-22-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00010737
InChI Key FHKSXSQHXQEMOK-LURJTMIESA-N
Synonym 1,2-hexanediol, 1,2-dihydroxyhexane, dl-1,2-hexanediol, 1,2-hexane diol, dl-hexane-1,2-diol, 5,6-dihydroxyhexane, 1,2-hexylene glycol, acmc-209o6j, 2,2,6,6-tetrakis hydroxymet cyclohexanol
PubChem CID 94335
IUPAC Name hexane-1,2-diol
SMILES CCCC[C@H](O)CO

Specifications

Density 0.951
Boiling Point 223°C to 224°C
Flash Point 122°C (251°F)
Refractive Index 1.442
Quantity 250 g
Beilstein 1719244
Sensitivity Hygroscopic
Solubility Information Miscible with water,lower aliphatic hydrocarbons and fatty acids.
Formula Weight 118.18
Percent Purity 97%
Chemical Name or Material 1,2-Hexanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.