missing translation for 'onlineSavingsMsg'
Learn More

1-(2-hydroxy-5-methoxyphenyl)ethan-1-one, ACROS Organics™

Product Code. 10245514
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10245514 25 g 25g
10045282 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10245514 Supplier Thermo Scientific Maybridge Supplier No. JFD00869EB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 705-15-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00008731
InChI Key MLIBGOFSXXWRIY-UHFFFAOYSA-N
Synonym 2'-hydroxy-5'-methoxyacetophenone, 1-2-hydroxy-5-methoxyphenyl ethanone, 2-hydroxy-5-methoxyacetophenone, 1-2-hydroxy-5-methoxyphenyl ethan-1-one, ethanone, 1-2-hydroxy-5-methoxyphenyl, 5-methoxy-2-hydroxyacetophenone, acetophenone, 2'-hydroxy-5'-methoxy, 2'-hydroxy-5'-methoxyacetylphenone, 1-2-hydroxy-5-methoxy-phenyl-ethanone, 1-acetyl-2-hydroxy-5-methoxybenzene
PubChem CID 69714
IUPAC Name 1-(2-hydroxy-5-methoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)OC)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 1-(2-Hydroxy-5-methoxyphenyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.