missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Hydroxyethyl)piperazine, 98+%

Product Code. 11493437
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
Product Code. Quantity unitSize
11493437 500 g 500g
11488780 10 g 10g
11483437 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11493437

Brand: Thermo Scientific Alfa Aesar A17215.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-76-4 | C6H14N2O | 130.191 g/mol

1-Piperazineethanol is a hydroxyalkyl substituted pieprazine used in the preparation of pharmaceutical compounds. It is also used as an Intermediate in the manufacture of polyurethane catalysts, corrosion inhibitors, surfactants, synthetic fibers, pharmaceuticals. It is also used to synthesize triethylenediamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Piperazineethanol is a hydroxyalkyl substituted pieprazine used in the preparation of pharmaceutical compounds. It is also used as an Intermediate in the manufacture of polyurethane catalysts, corrosion inhibitors, surfactants, synthetic fibers, pharmaceuticals. It is also used to synthesize triethylenediamine.

Solubility
Miscible with water. Also miscible in ethanol, acetone, ether benzene. Immiscible in hexane.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-76-4
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.191
MDL Number MFCD00005970
InChI Key WFCSWCVEJLETKA-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl piperazine, 1-piperazineethanol, 1-2-hydroxyethyl piperazine, hydroxyethylpiperazine, 2-1-piperazinyl ethanol, 2-hydroxyethylpiperazine, 2-piperazinoethanol, 2-piperazin-1-yl ethan-1-ol, 1-piperazinethanol, 4-2-hydroxyethyl piperazine
PubChem CID 7677
IUPAC Name 2-piperazin-1-ylethanol
SMILES C1CN(CCN1)CCO

Specifications

Melting Point -39°C
Density 1.06
pH 12.3
Boiling Point 242°C to 246°C
Flash Point 135°C (275°F)
Odor Amine-like
Refractive Index 1.5065
Quantity 500 g
Beilstein 104361
Solubility Information Miscible with water. Also miscible in ethanol,acetone,ether benzene. Immiscible in hexane.
Formula Weight 130.19
Percent Purity ≥98%
Chemical Name or Material 1-(2-Hydroxyethyl)piperazine
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.