missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Hydroxyethyl)piperidine, 99%

Product Code. 11400288
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400288 500 g 500g
11490278 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400288 Supplier Thermo Scientific Alfa Aesar Supplier No. B23816.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3040-44-6 | C7H15NO | 129.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3040-44-6
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00006512
InChI Key KZTWONRVIPPDKH-UHFFFAOYSA-N
Synonym 2-piperidinoethanol, 1-piperidineethanol, 2-piperidin-1-yl ethanol, 1-2-hydroxyethyl piperidine, n-2-hydroxyethyl piperidine, n-piperidineethanol, 2-1-piperidinyl ethanol, n-hydroxyethyl piperidine, beta-piperidylethanol, beta-piperidinoethanol
PubChem CID 18232
ChEBI CHEBI:61238
IUPAC Name 2-piperidin-1-ylethanol
SMILES C1CCN(CC1)CCO

Specifications

Melting Point 16°C
Density 0.977
pH 12.4
Boiling Point 199°C to 202°C
Flash Point 83°C (181°F)
Odor Amine-like
Refractive Index 1.4804
Quantity 500 g
UN Number UN2735
Beilstein 103390
Sensitivity Air Sensitive
Formula Weight 129.2
Percent Purity 99%
Chemical Name or Material 1-(2-Hydroxyethyl)piperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.