missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-1,2-Isopropylidineglycerol-1,1,2,3,3-d5, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | (R,S)-2,2-Dimethyl-1,3-dioxolane-4-methanol-d5 |
| CAS | 74300-14-4 |
| Molecular Formula | C6 D5 H7 O3 |
| Quantity | 0.25 g |
| Synonym | (RS)-Solketal-d5, (+/-)-1,2-O-Isopropylideneglycerol-d5, (+/-)-Glycerol 1,2-Acetonide-d5, 1,2-Isopropylideneglycerin-d5, 1,2-Isopropylideneglycerol-d5, 2,3-(Isopropylidenedioxy)propanol-d5, 2,3-Isopropylideneglycerol-d5, Acetone Glyceryl Acetal-d5, Dioxolan-d5, α,β-Isopropylidene-d5 ether, Glycerolacetone-d5, NSC 59720-d5; |
| InChI Formula | InChI=1 S/C6H12O3/c1-6(2)8-4-5(3-7)9-6/h5,7 H,3-4H2,1-2H3/i3D2,4D2,5 D |
| SMILES | [2 H]C([2 H])(O)C1([2 H])OC(C)(C)OC1([2 H])[2 H] |
| IUPAC Name | dideuterio-(4,5,5-trideuterio-2,2-dimethyl-1,3-dioxolan-4-yl)methanol |
| Molecular Weight (g/mol) | 137.19 |
| Formula Weight | 137.11 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction