missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methoxyphenethyl)piperazine, TRC

Product Code. 30477754
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477754 10 g 10g
30483018 5 g 5g
30478970 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477754 Supplier Toronto Research Chemicals Supplier No. TRCM21983810G
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6722-54-9
Molecular Formula C13H20N2O
Molecular Weight (g/mol) 220.32
Synonym 1-(2-Methoxy-2-Phenylethyl)Piperazine
IUPAC Name 1-(2-methoxy-2-phenylethyl)piperazine
SMILES COC(CN1CCNCC1)c2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H20N2O/c1-16-13(12-5-3-2-4-6-12)11-15-9-7-14-8-10-15/h2-6,13-14H,7-11H2,1H3
Formula Weight 220.1576
Chemical Name or Material 1-(2-Methoxyphenethyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.