missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methoxyphenyl)piperazine, 98%, Thermo Scientific™

Product Code. 11462488
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462488 5 g 5g
11472488 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11462488 Supplier Thermo Scientific Alfa Aesar Supplier No. A15819.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate. N-alkylated 1-(2-methyoxyphenyl)piperazines markedly improved affinity and selectivity of the dopamine D3 receptor which is recognized as a potential therapeutic target for the treatment of various neurological and psychiatric disorders.

Solubility
Soluble in chloroform, ethyl acetate, and methanol. Insoluble in water

Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35386-24-4
Molecular Formula C11H18Cl2N2O
Molecular Weight (g/mol) 265.18
MDL Number MFCD00005958
InChI Key ZGWQDMTYAQEMHA-UHFFFAOYSA-N
Synonym 1-2-methoxyphenyl piperazine, 1-2-methoxy-phenyl-piperazine, 1-o-methoxyphenyl piperazine, 2-mpp, 1-2-methoxyphenyl-piperazine, n-2-methoxyphenyl piperazine, unii-81njo1330a, 1-2-methoxy phenyl piperazine, 2-methoxyphenylpiperazine, piperazine, 1-2-methoxyphenyl
PubChem CID 1346
SMILES [H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1N1CCNCC1

Specifications

Melting Point 35°C to 39°C
Density 1.106
Boiling Point 130°C to 133°C (0.1 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN2923
Beilstein 167888
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,ethyl acetate,and methanol. Insoluble in water
IUPAC Name 1-(2-methoxyphenyl)piperazine
Formula Weight 192.26
Percent Purity 98%
Chemical Name or Material 1-(2-Methoxyphenyl)piperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.