missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methylphenyl)ethanamine, TRC

Product Code. 30478083
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478083 500 mg 500mg
30468912 50 mg 50mg
30480843 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478083 Supplier Toronto Research Chemicals Supplier No. TRCM339498500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42142-17-6
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
SMILES CC(N)c1ccccc1C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H13N/c1-7-5-3-4-6-9(7)8(2)10/h3-6,8H,10H2,1-2H3
Formula Weight 135.1048
Chemical Name or Material 1-(2-Methylphenyl)ethanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.