missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2-O-Isopropylidene-alpha-D-xylofuranose, 98%

Product Code. 15487349
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487349 25 g 25g
15467349 5 g 5g
15477349 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15487349 Supplier Thermo Scientific Chemicals Supplier No. J66110.14
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 20031-21-4 | C8H14O5 | 190.195 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20031-21-4
Molecular Formula C8H14O5
Molecular Weight (g/mol) 190.195
MDL Number MFCD00063295
InChI Key JAUQZVBVVJJRKM-XZBKPIIZSA-N
Synonym 1,2-o-isopropylidene-alpha-d-xylofuranose, 3ar,5r,6s,6ar-5-hydroxymethyl-2,2-dimethyltetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2-o-isopropylidene-a-d-xylofuranose, alpha-d-xylofuranose, 1,2-o-1-methylethylidene, 1,2-o-isopropylidene-d-xylofuranose, 3ar,5r,6s,6ar-5-hydroxymethyl-2,2-dimethyl-tetrahydro-2h-furo 2,3-d 1,3 dioxol-6-ol, 3ar,5r,6s,6ar-5-hydroxymethyl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 3ar,5r,6s,6ar-5-hydroxymethyl-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, .alpha.-d-xylofuranose, 1,2-o-1-methylethylidene, 1,2-o-isopropylidene-, a-d-xylofuranose
PubChem CID 88338
IUPAC Name (3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
SMILES CC1(OC2C(C(OC2O1)CO)O)C

Specifications

Melting Point 69°C to 71°C
Color White
Quantity 25 g
Physical Form Powder
Beilstein 81520
Boiling Point 112°C to 114°C (0.06 mmHg)
Formula Weight 190.19
Percent Purity 98%
Chemical Name or Material 1,2-O-Isopropylidene-alpha-D-xylofuranose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.