missing translation for 'onlineSavingsMsg'
Learn More

1-(2-(Phenanthren-9-yl)phenyl)propan-2-one, TRC

Product Code. 30424334
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424334 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424334 Supplier Toronto Research Chemicals Supplier No. TRCP31252050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(2-(Phenanthren-9-yl)phenyl)propan-2-one
CAS 1013933-56-6
Molecular Formula C23H18O
Quantity 50 mg
InChI Formula InChI=1S/C23H18O/c1-16(24)14-17-8-2-5-11-20(17)23-15-18-9-3-4-10-19(18)21-12-6-7-13-22(21)23/h2-13,15H,14H2,1H3
SMILES CC(=O)Cc1ccccc1c2cc3ccccc3c4ccccc24
IUPAC Name 1-(2-phenanthren-9-ylphenyl)propan-2-one
Molecular Weight (g/mol) 310.39
Formula Weight 310.1358
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.