missing translation for 'onlineSavingsMsg'
Learn More

1,2-Phenylenediacetic acid, 99%

Product Code. 10734641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10734641 5 g Glass Bottle 5g
10582171 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10734641 Supplier Thermo Scientific Acros Supplier No. 157550050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7500-53-0 | C10H10O4 | 194.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7500-53-0
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00004329
InChI Key MMEDJBFVJUFIDD-UHFFFAOYSA-N
Synonym 1,2-phenylenediacetic acid, 2,2'-1,2-phenylene diacetic acid, o-phenylenediacetic acid, o-benzenediacetic acid, 1,2-benzenediacetic acid, 2-2-carboxymethyl phenyl acetic acid, phenylenediacetic acid, acmc-209owx, 1,2-benzenediaceticacid, o-phenylene-diacetic acid
PubChem CID 24123
IUPAC Name 2-[2-(carboxymethyl)phenyl]acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)CC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 150°C to 152°C
Color Beige to Beige-Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula C6H4(CH2CO2H)2
Quantity 5 g
Beilstein 09, 874
Formula Weight 194.19
Percent Purity 99%
Physical Form Needle-like Crystalline Powder
Chemical Name or Material 1, 2-Phenylenediacetic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Phenylenediacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.